4,6-Dibromoisophthalic acid 97%

Supplier: Ambeed
Warning

A1275262-100MG A1275262-250MG A1275262-5G A1275262-1G
76681-382EA 26 USD
76681-382 76681-384 76681-386 77592-484
4,6-Dibromoisophthalic acid 97%
4,6-Dibromoisophthalic acid

Precaución: for Research Use Only

Formula: C₈H₄Br₂O₄
MDL Number: MFCD32641193
CAS Number: 24063-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR