N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-6-phenyl-5,6,6a,7,8,9,10,10a-octahydro-7,10-methanophenanthridine-2-carboxamide 97%

Supplier: Ambeed
Warning

A1275164-100MG A1275164-250MG A1275164-50MG
76681-374EA 153.84 USD
76681-374 76681-376 76681-378
N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-6-phenyl-5,6,6a,7,8,9,10,10a-octahydro-7,10-methanophenanthridine-2-carboxamide 97%
N-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-6-phenyl-5,6,6a,7,8,9,10,10a-octahydro-7,10-methanophenanthridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₉H₂₈N₂O₃
MW: 452.5442 g/mol
MDL Number: MFCD04215212
CAS Number: 1005268-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR