5-((3AS,8aS)-2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)-5H-dibenzo[b,f]azepine 98%

Supplier: Ambeed
Warning

A1274634-100MG A1274634-1G A1274634-250MG
76681-360EA 100.79 USD
76681-360 76681-362 76681-364
5-((3AS,8aS)-2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)-5H-dibenzo[b,f]azepine 98%
5-((3AS,8aS)-2,2-Dimethyl-4,4,8,8-tetraphenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepin-6-yl)-5H-dibenzo[b,f]azepine

Precaución: for Research Use Only

Formula: C₄₅H₃₈NO₄P
MW: 687.7613 g/mol
CAS Number: 2411176-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR