(11AS)-5-(4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-yl)-5H-dibenzo[b,f]azepine 98%

Supplier: Ambeed
Warning

A1274633-100MG A1274633-250MG
76681-356EA 125.19 USD
76681-356 76681-358
(11AS)-5-(4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-yl)-5H-dibenzo[b,f]azepine 98%
(11AS)-5-(4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-12-yl)-5H-dibenzo[b,f]azepine

Precaución: for Research Use Only

Formula: C₃₁H₂₄NO₂P
MW: 473.5 g/mol
CAS Number: 1361055-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR