5-(Aminomethyl)-N-methylpyridin-2-amine 98%

Supplier: Ambeed
Danger

A1274469-100MG A1274469-1G A1274469-250MG
76681-332EA 82.76 USD
76681-332 76681-334 76681-336
5-(Aminomethyl)-N-methylpyridin-2-amine 98%
5-(Aminomethyl)-N-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃
MW: 137.1823 g/mol
MDL Number: MFCD12403499
CAS Number: 779324-37-7
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR