Methyl 2-chloro-4-phenoxybenzoate 98%

Supplier: Ambeed
Warning

A1273394-1G A1273394-250MG A1273394-5G
76681-308EA 10 USD
76681-308 76681-310 76681-312
Methyl 2-chloro-4-phenoxybenzoate 98%
Methyl 2-chloro-4-phenoxybenzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₃
MW: 262.6883 g/mol
MDL Number: MFCD34181685
CAS Number: 2095393-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR