N,N-Dimethyl-5,6,7,8-tetrahydrobenzo[4,5]thieno[2,3-d]pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A1273189-100MG A1273189-1G A1273189-250MG
76681-288EA 62.6 USD
76681-288 76681-290 76681-292
N,N-Dimethyl-5,6,7,8-tetrahydrobenzo[4,5]thieno[2,3-d]pyrimidin-4-amine 97%
N,N-Dimethyl-5,6,7,8-tetrahydrobenzo[4,5]thieno[2,3-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃S
MW: 233.3326 g/mol
MDL Number: MFCD01192168
CAS Number: 871807-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR