1-(1H-Indol-6-yl)cyclopropanol 97%

Supplier: Ambeed
Warning

A1273163-1G A1273163-250MG
76681-282EA 335.25 USD
76681-282 76681-284
1-(1H-Indol-6-yl)cyclopropanol 97%
1-(1H-Indol-6-yl)cyclopropanol

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO
MW: 173.2111 g/mol
MDL Number: MFCD28357582
CAS Number: 1891321-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR