9-Bromo-2,3,4,5-tetrahydrobenzo[b]oxepine 97%

Supplier: Ambeed
Warning

A1273109-100MG A1273109-1G A1273109-250MG
76681-266EA 75.33 USD
76681-266 76681-268 76681-270
9-Bromo-2,3,4,5-tetrahydrobenzo[b]oxepine 97%
9-Bromo-2,3,4,5-tetrahydrobenzo[b]oxepine

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
MW: 227.0977 g/mol
MDL Number: MFCD22035661
CAS Number: 1369140-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR