Dichloro(3,4,7,8-tetramethyl-1,10-phenanthroline-κN1,κN10)-Nickel 98%

Supplier: Ambeed
Danger

A1272894-100MG A1272894-1G A1272894-250MG A1272894-5G
76681-258EA 45.62 USD
76681-258 76681-260 76681-262 77621-426
Dichloro(3,4,7,8-tetramethyl-1,10-phenanthroline-κN1,κN10)-Nickel 98%
Dichloro(3,4,7,8-tetramethyl-1,10-phenanthroline-κN1,κN10)-Nickel

Precaución: for Research Use Only

Formula: C₁₆H₁₆Cl₂N₂Ni
MW: 365.9110 g/mol
CAS Number: 2254430-88-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR