(T-4)-Dichloro(1,10-phenanthroline-κN1,κN10)nickel 98%

Supplier: Ambeed
Danger

A1272892-1G A1272892-25G A1272892-5G
76681-254EA 29.71 USD
76681-254 76681-256 77621-422
(T-4)-Dichloro(1,10-phenanthroline-κN1,κN10)nickel 98%
(T-4)-Dichloro(1,10-phenanthroline-κN1,κN10)nickel

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂N₂Ni
MW: 309.8047 g/mol
CAS Number: 22980-76-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR