(S)-2-(4-Methoxy-6-methylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1272887-100MG A1272887-1G A1272887-250MG
76681-238EA 149 USD
76681-238 76681-240 76681-242
(S)-2-(4-Methoxy-6-methylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
(S)-2-(4-Methoxy-6-methylpyridin-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₂
CAS Number: 2757082-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR