(S)-4-(tert-Butyl)-2-(4-methoxy-6-methylpyridin-2-yl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A1272886-100MG A1272886-1G A1272886-250MG
76681-232EA 159.14 USD
76681-232 76681-234 76681-236
(S)-4-(tert-Butyl)-2-(4-methoxy-6-methylpyridin-2-yl)-4,5-dihydrooxazole 98%
(S)-4-(tert-Butyl)-2-(4-methoxy-6-methylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.3208 g/mol
CAS Number: 235742-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR