(R)-4-(tert-Butyl)-2-(4-methoxy-6-methylpyridin-2-yl)-4,5-dihydrooxazole 95%

Supplier: Ambeed
Warning

A1272864-100MG A1272864-1G A1272864-250MG
76681-220EA 122.01 USD
76681-220 76681-222 76681-224
(R)-4-(tert-Butyl)-2-(4-methoxy-6-methylpyridin-2-yl)-4,5-dihydrooxazole 95%
(R)-4-(tert-Butyl)-2-(4-methoxy-6-methylpyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₂
MW: 248.3208 g/mol
CAS Number: 2757082-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR