4-Iodo-2-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A1272457-1G A1272457-250MG A1272457-5G
76681-214EA 70.02 USD
76681-214 76681-216 76681-218
4-Iodo-2-methylbenzonitrile 98%
4-Iodo-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆IN
MW: 243.0444 g/mol
MDL Number: MFCD18397801
CAS Number: 52107-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR