2-Chloro-3-fluoro-5-iodopyridin-4-amine 98%

Supplier: Ambeed
Warning

A1272011-250MG A1272011-1G A1272011-100MG
76681-212EA 25 USD
76681-212 77592-472 77596-642
2-Chloro-3-fluoro-5-iodopyridin-4-amine 98%
2-Chloro-3-fluoro-5-iodopyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₃ClFIN₂
MDL Number: MFCD31672865
CAS Number: 1801841-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR