4,4',4'',4'''-(9H-Carbazole-1,3,6,8-tetrayl)tetrabenzoic acid 98%

Supplier: Ambeed
Warning

A1271744-100MG A1271744-1G A1271744-250MG
76681-200EA 39.26 USD
76681-200 76681-202 76681-204
4,4',4'',4'''-(9H-Carbazole-1,3,6,8-tetrayl)tetrabenzoic acid 98%
4,4',4'',4'''-(9H-Carbazole-1,3,6,8-tetrayl)tetrabenzoic acid

Precaución: for Research Use Only

Formula: C₄₀H₂₅NO₈
MW: 647.6284 g/mol
CAS Number: 2097131-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR