4,4',4''-(Pyridine-2,4,6-triyl)tribenzoic acid 98%

Supplier: Ambeed
Warning

A1271742-100MG A1271742-1G A1271742-250MG
76681-194EA 23 USD
76681-194 76681-196 76681-198
4,4',4''-(Pyridine-2,4,6-triyl)tribenzoic acid 98%
4,4',4''-(Pyridine-2,4,6-triyl)tribenzoic acid

Precaución: for Research Use Only

Formula: C₂₆H₁₇NO₆
MDL Number: MFCD32856651
CAS Number: 107063-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR