4,4'-(Anthracene-9,10-diylbis(ethyne-2,1-diyl))dibenzaldehyde 98%

Supplier: Ambeed
Warning

A1271741-100MG A1271741-1G A1271741-250MG
76681-188EA 25.47 USD
76681-188 76681-190 76681-192
4,4'-(Anthracene-9,10-diylbis(ethyne-2,1-diyl))dibenzaldehyde 98%
4,4'-(Anthracene-9,10-diylbis(ethyne-2,1-diyl))dibenzaldehyde

Precaución: for Research Use Only

Formula: C₃₂H₁₈O₂
MW: 434.4841 g/mol
CAS Number: 1361119-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR