Diethyl 4,4'-(ethyne-1,2-diyl)dibenzoate 98%

Supplier: Ambeed
Warning

A1271739-250MG A1271739-1G A1271739-5G
76681-186EA 29.71 USD
76681-186 77592-470 77621-412
Diethyl 4,4'-(ethyne-1,2-diyl)dibenzoate 98%
Diethyl 4,4'-(ethyne-1,2-diyl)dibenzoate

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₄
MW: 322.3545 g/mol
CAS Number: 83536-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR