5',5''''-([2,2'-Bithiophene]-5,5'-diyl)bis(([1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid)) 98%

Supplier: Ambeed
Warning

A1271736-100MG A1271736-1G A1271736-250MG
76681-180EA 64.72 USD
76681-180 76681-182 76681-184
5',5''''-([2,2'-Bithiophene]-5,5'-diyl)bis(([1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid)) 98%
5',5''''-([2,2'-Bithiophene]-5,5'-diyl)bis(([1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid))

Precaución: for Research Use Only

Formula: C₄₈H₃₀O₈S₂
MW: 798.8770 g/mol
CAS Number: 1227780-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR