((1,3,5-Triazine-2,4,6-triyl)tris(benzene-4,1-diyl))triboronic acid 98%

Supplier: Ambeed
Warning

A1271735-100MG A1271735-1G A1271735-250MG
76681-174EA 48.81 USD
76681-174 76681-176 76681-178
((1,3,5-Triazine-2,4,6-triyl)tris(benzene-4,1-diyl))triboronic acid 98%
((1,3,5-Triazine-2,4,6-triyl)tris(benzene-4,1-diyl))triboronic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₈B₃N₃O₆
MW: 440.8171 g/mol
CAS Number: 910231-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR