4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-3,5-dicarbaldehyde)) 98%

Supplier: Ambeed
Warning

A1271732-100MG A1271732-1G A1271732-250MG
76681-168EA 18 USD
76681-168 76681-170 76681-172
4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-3,5-dicarbaldehyde)) 98%
4',4''',4'''''-Nitrilotris(([1,1'-biphenyl]-3,5-dicarbaldehyde))

Precaución: for Research Use Only

Formula: C₄₂H₂₇NO₆
MW: 641.6669 g/mol
CAS Number: 2030105-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR