5,5'-(Ethyne-1,2-diyl)diisophthalaldehyde 98%

Supplier: Ambeed
Warning

A1271728-100MG A1271728-250MG A1271728-1G
76681-164EA 54.11 USD
76681-164 76681-166 77592-468
5,5'-(Ethyne-1,2-diyl)diisophthalaldehyde 98%
5,5'-(Ethyne-1,2-diyl)diisophthalaldehyde

Precaución: for Research Use Only

Formula: C₁₈H₁₀O₄
MW: 290.2696 g/mol
MDL Number: MFCD34167503
CAS Number: 2237965-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR