(R)-N-((S)-(2-(dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 98%

Supplier: Ambeed
Warning

A1271286-100MG A1271286-250MG A1271286-50MG
76681-146EA 165 USD
76681-146 76681-148 76681-150
(R)-N-((S)-(2-(dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide 98%
(R)-N-((S)-(2-(dicyclohexylphosphanyl)phenyl)(phenyl)methyl)-N,2-dimethylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₀H₄₄NOPS
MDL Number: MFCD32697196
CAS Number: 2241598-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR