(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 98%

Supplier: Ambeed
Warning

Synonyms: (2S,5S)-(-)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane, 2-{2-[(2S,5S)-2,5-Diethylphospholan-1-yl]phenyl}-1,3-dioxolane, 2-{2-[(2S,5S)-2,5-Diethyl-1-phospholano]phenyl}1,3-dioxolane

A1271223-100MG A1271223-1G A1271223-250MG
76681-140EA 54.11 USD
76681-140 76681-142 76681-144
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane 98%
(2S,5S)-1-(2-(1,3-Dioxolan-2-yl)phenyl)-2,5-diethylphospholane

Precaución: for Research Use Only

Formula: C₁₇H₂₅O₂P
MW: 292.36 g/mol
MDL Number: MFCD09265099
CAS Number: 1314246-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR