4,6-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A1271155-250MG A1271155-1G
76681-130EA 209 USD
76681-130 77596-640
4,6-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 97%
4,6-Bis((S)-4-benzyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₃₂H₂₆N₂O₃
MW: 486.5605 g/mol
CAS Number: 2126892-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR