4,6-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A1271153-100MG A1271153-1G A1271153-250MG
76681-124EA 165.51 USD
76681-124 76681-126 76681-128
4,6-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan 97%
4,6-Bis((S)-4-(tert-butyl)-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₂₆H₃₀N₂O₃
MW: 418.5300 g/mol
CAS Number: 2067322-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR