(2S,6S)-4-(tert-Butoxycarbonyl)-6-methylmorpholine-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A1271135-250MG A1271135-1G
76681-122EA 123 USD
76681-122 77867-132
(2S,6S)-4-(tert-Butoxycarbonyl)-6-methylmorpholine-2-carboxylic acid 96%
(2S,6S)-4-(tert-Butoxycarbonyl)-6-methylmorpholine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
CAS Number: 2165414-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR