3-((1H-Benzo[d]imidazol-2-yl)amino)-4-(((S)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl)amino)cyclobut-3-ene-1,2-dione 97%

Supplier: Ambeed
Warning

A1271072-100MG A1271072-250MG
76681-118EA 100.79 USD
76681-118 76681-120
3-((1H-Benzo[d]imidazol-2-yl)amino)-4-(((S)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl)amino)cyclobut-3-ene-1,2-dione 97%
3-((1H-Benzo[d]imidazol-2-yl)amino)-4-(((S)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methyl)amino)cyclobut-3-ene-1,2-dione

Precaución: for Research Use Only

Formula: C₃₁H₃₀N₆O₃
MW: 534.61 g/mol
CAS Number: 2565792-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR