(11bS)-2,6-Bis(4-(tert-butyl)phenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%

Supplier: Ambeed
Warning

A1270816-100MG A1270816-1G A1270816-250MG
76681-112EA 101.85 USD
76681-112 76681-114 76681-116
(11bS)-2,6-Bis(4-(tert-butyl)phenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide 98%
(11bS)-2,6-Bis(4-(tert-butyl)phenyl)-4-hydroxy-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₄₀H₄₅O₄P
MW: 620.7567 g/mol
CAS Number: 2757287-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR