(S)-3,3'-Di([1,1'-biphenyl]-4-yl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 97%

Supplier: Ambeed
Warning

A1270801-100MG A1270801-1G A1270801-250MG
76681-082EA 111.4 USD
76681-082 76681-084 76681-086
(S)-3,3'-Di([1,1'-biphenyl]-4-yl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol 97%
(S)-3,3'-Di([1,1'-biphenyl]-4-yl)-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₄₄H₃₈O₂
MW: 598.7713 g/mol
CAS Number: 2757287-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR