2-Mesityl-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1270754-50MG A1270754-100MG A1270754-250MG A1270754-1G
76681-074EA 35 USD
76681-074 77621-398 77621-400 77867-128
2-Mesityl-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate 97%
2-Mesityl-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₄H₁₈BF₄N₃O
MDL Number: MFCD32640817
CAS Number: 2170152-85-7
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR