(R)-1-Tosylazetidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A1270657-100MG A1270657-250MG
76681-064EA 13.8 USD
76681-064 76681-066
(R)-1-Tosylazetidine-2-carboxylic acid 97%
(R)-1-Tosylazetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄S
MW: 255.2902 g/mol
MDL Number: MFCD06795485
CAS Number: 174740-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR