Alizarin red S (sodium salt) indicator

Supplier: Ambeed
Warning

Synonyms: Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium

A1270627-25G A1270627-100G A1270627-500G
76681-054EA 22 USD
76681-054 77596-638 77867-126
Alizarin red S (sodium salt) indicator
Alizarin Red S (sodium salt)

Precaución: for Research Use Only

Formula: C₁₄H₇NaO₇S
MW: 342.26 g/mol
MDL Number: MFCD00013049
CAS Number: 130-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR