Lithium 4,5-dicyano-2-(trifluoromethyl)imidazol-1-ide 97%

Supplier: Ambeed
Danger

A1264680-5G A1264680-25G
76681-046EA 169.75 USD
76681-046 77566-454
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazol-1-ide 97%
Lithium 4,5-dicyano-2-(trifluoromethyl)imidazol-1-ide

Precaución: For research use only.

Formula: C₆F₃LiN₄
MW: 192.0272 g/mol
MDL Number: MFCD18827480
CAS Number: 761441-54-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR