7-Chloro-1-(4-(trifluoromethyl)benzyl)indoline-2,3-dione 98%

Supplier: Ambeed
Warning

Synonyms: VU0238441

A1264493-10MG A1264493-1MG A1264493-25MG A1264493-50MG A1264493-5MG
76681-016EA 41.38 USD
76681-016 76681-018 76681-020 76681-022 76681-024
7-Chloro-1-(4-(trifluoromethyl)benzyl)indoline-2,3-dione 98%
7-Chloro-1-(4-(trifluoromethyl)benzyl)indoline-2,3-dione

Precaución: for Research Use Only

Formula: C₁₆H₉ClF₃NO₂
MW: 339.6964 g/mol
MDL Number: MFCD18642601
CAS Number: 85511-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR