(S)-3-(4-Acrylamidobenzamido)-N-(2-(dimethylamino)-1-phenylethyl)-6,6-dimethyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: YKL-5-124

A1264490-25MG A1264490-10MG A1264490-50MG
76681-014EA 232.34 USD
76681-014 77621-388 77621-390
(S)-3-(4-Acrylamidobenzamido)-N-(2-(dimethylamino)-1-phenylethyl)-6,6-dimethyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide 98%
(S)-3-(4-Acrylamidobenzamido)-N-(2-(dimethylamino)-1-phenylethyl)-6,6-dimethyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide

Precaución: for Research Use Only

Formula: C₂₈H₃₃N₇O₃
MW: 515.61 g/mol
MDL Number: MFCD34469131
CAS Number: 1957203-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR