(3aS,3a'S,7aS,7a'S)-2,2'-(((2S,4S)-Pentane-2,4-diyl)bis(oxy))bis(1,3-diphenyloctahydro-1H-benzo[d][1,3,2]diazaphosphole) 98%
Supplier: Ambeed
|
Warning
|
A1264392-100MG
A1264392-1G
A1264392-250MG
76681-008EA
96.55
USD
76681-008
76681-010
76681-012
(3aS,3a'S,7aS,7a'S)-2,2'-(((2S,4S)-Pentane-2,4-diyl)bis(oxy))bis(1,3-diphenyloctahydro-1H-benzo[d][1,3,2]diazaphosphole) 98%
(3aS,3a'S,7aS,7a'S)-2,2'-(((2S,4S)-Pentane-2,4-diyl)bis(oxy))bis(1,3-diphenyloctahydro-1H-benzo[d][1,3,2]diazaphosphole)
Precaución: for Research Use Only
Formula:
C₄₁H₅₀N₄O₂P₂ MW: 692.8088 g/mol |
CAS Number:
1384619-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...