(3aR,3a'R,7aR,7a'R)-2,2'-(((2S,4S)-Pentane-2,4-diyl)bis(oxy))bis(1,3-diphenyloctahydro-1H-benzo[d][1,3,2]diazaphosphole) 98%

Supplier: Ambeed
Warning

A1264391-100MG A1264391-1G A1264391-250MG
76681-002EA 145.35 USD
76681-002 76681-004 76681-006
(3aR,3a'R,7aR,7a'R)-2,2'-(((2S,4S)-Pentane-2,4-diyl)bis(oxy))bis(1,3-diphenyloctahydro-1H-benzo[d][1,3,2]diazaphosphole) 98%
(3aR,3a'R,7aR,7a'R)-2,2'-(((2S,4S)-Pentane-2,4-diyl)bis(oxy))bis(1,3-diphenyloctahydro-1H-benzo[d][1,3,2]diazaphosphole)

Precaución: for Research Use Only

Formula: C₄₁H₅₀N₄O₂P₂
MW: 692.8088 g/mol
CAS Number: 2370958-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR