(4R,4'R)-2,2'-Methylenebis[4,5-dihydro-4-(1-methylethyl)oxazole] 95%

Supplier: Ambeed
Warning

A1264384-100MG A1264384-250MG A1264384-1G
76680-970EA 29.71 USD
76680-970 76680-972 77621-384
(4R,4'R)-2,2'-Methylenebis[4,5-dihydro-4-(1-methylethyl)oxazole] 95%
(4R,4'R)-2,2'-Methylenebis[4,5-dihydro-4-(1-methylethyl)oxazole]

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₂
MW: 238.3260 g/mol
CAS Number: 164976-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR