H-Trp(Boc)-OH 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)propanoic acid

A1263605-1G A1263605-5G A1263605-25G
76680-938EA 16.98 USD
76680-938 76680-940 77571-566
H-Trp(Boc)-OH 97%
H-Trp(Boc)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₄
MW: 304.346 g/mol
MDL Number: MFCD00237294
CAS Number: 146645-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR