PEG5-Tos 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tos-PEG5-OH, 14-Hydroxy-3,6,9,12-tetraoxatetradecyl 4-methylbenzenesulphonate
A1263601-100MG
A1263601-1G
A1263601-250MG
A1263601-5G
76680-930EA
38.2
USD
76680-930
76680-932
76680-934
76680-936
PEG5-Tos 98%
PEG5-Tos
Precaución: for Research Use Only
Formula:
C₁₇H₂₈O₈S MW: 392.46 g/mol |
MDL Number:
MFCD22574780 CAS Number: 155130-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...