(R)-N-((S)-(2-(diphenylphosphaneyl)phenyl)(naphthalen-1-yl)methyl)-2-methylpropane-2-sulfinamide 98%

Supplier: Ambeed
Warning

A1263576-250MG A1263576-50MG A1263576-100MG
76680-926EA 301 USD
76680-926 76680-928 77621-372
(R)-N-((S)-(2-(diphenylphosphaneyl)phenyl)(naphthalen-1-yl)methyl)-2-methylpropane-2-sulfinamide 98%
(R)-N-((S)-(2-(diphenylphosphaneyl)phenyl)(naphthalen-1-yl)methyl)-2-methylpropane-2-sulfinamide

Precaución: for Research Use Only

Formula: C₃₃H₃₂NOPS
MDL Number: MFCD32697147
CAS Number: 1595319-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR