Methyl 2-chloro-6-formylnicotinate 97%

Supplier: Ambeed
Danger

A1263374-1G A1263374-250MG
76680-916EA 434.97 USD
76680-916 76680-918
Methyl 2-chloro-6-formylnicotinate 97%
Methyl 2-chloro-6-formylnicotinate

Precaución: for Research Use Only

Formula: C₈H₆ClNO₃
MW: 199.5911 g/mol
MDL Number: MFCD17676620
CAS Number: 1263286-09-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR