(S)-4-Methyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole 98%
Supplier: Ambeed
|
Warning
|
A1263095-100MG
A1263095-1G
A1263095-250MG
76680-896EA
51
USD
76680-896
76680-898
76680-900
(S)-4-Methyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole 98%
(S)-4-Methyl-2-(5-(trifluoromethyl)pyridin-2-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃N₂O MW: 230.1865 g/mol |
CAS Number:
1803416-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...