3-Chloro-1,1-diethoxypropane 95% stabilised

Supplier: Ambeed
Danger

Synonyms: 3-Chloropropionaldehydediethylacetal, 3-Chloropropionaldehyde diethyl acetal

Stabilization: Stabilised with potassium carbonate

A1263026-25G A1263026-5G
76680-890EA 31 USD
76680-890 76680-892
3-Chloro-1,1-diethoxypropane 95% stabilised
3-Chloro-1,1-diethoxypropane

Precaución: for Research Use Only

Formula: C₇H₁₅ClO₂
MDL Number: MFCD00000994
CAS Number: 35573-93-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR