3,3',4,5'-Tetramethoxypiceatannol 97%

Supplier: Ambeed
Warning

Synonyms: (E)-4-(3,5-Dimethoxystyryl)-1,2-dimethoxybenzene

A1262996-100MG A1262996-1G A1262996-250MG
76680-884EA 50.93 USD
76680-884 76680-886 76680-888
3,3',4,5'-Tetramethoxypiceatannol 97%
3,3',4,5'-Tetramethoxypiceatannol

Precaución: for Research Use Only

Formula: C₁₈H₂₀O₄
MW: 300.3490 g/mol
MDL Number: MFCD09033675
CAS Number: 83088-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR