Methyl 2-(2-iodo-3,4-dimethoxyphenyl)acetate 97%

Supplier: Ambeed
Warning

A1262892-1G A1262892-250MG
76680-866EA 370.26 USD
76680-866 76680-868
Methyl 2-(2-iodo-3,4-dimethoxyphenyl)acetate 97%
Methyl 2-(2-iodo-3,4-dimethoxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₃IO₄
MW: 336.1230 g/mol
MDL Number: MFCD28793599
CAS Number: 1806331-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR