(S)-8-(2,4-Dichlorophenyl)-9-(4-((1-(3-fluoropropyl)pyrrolidin-3-yl)oxy)phenyl)-6,7-dihydro-5H-benzo[7]annulene-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Amcenestrant

A1262263-10MG A1262263-25MG A1262263-50MG
76680-856EA 112.46 USD
76680-856 77621-366 77621-368
(S)-8-(2,4-Dichlorophenyl)-9-(4-((1-(3-fluoropropyl)pyrrolidin-3-yl)oxy)phenyl)-6,7-dihydro-5H-benzo[7]annulene-3-carboxylic acid 97%
(S)-8-(2,4-Dichlorophenyl)-9-(4-((1-(3-fluoropropyl)pyrrolidin-3-yl)oxy)phenyl)-6,7-dihydro-5H-benzo[7]annulene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₃₁H₃₀Cl₂FNO₃
MW: 554.48 g/mol
CAS Number: 2114339-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR